siRNA Details
| Gene Symbol: Ggnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153144.2 | NP_694784.2 | 4 | 404 | ||
| GenBank mRNA | AB064543.1 | - | 4 | 404 | ||
| AY633741.1 | - | 4 | 404 | |||
| BC060738.1 | - | 3 | 360 | |||
| BC139772.1 | - | 3 | 310 | 40111779 | ||
| BC145605.1 | - | 3 | 312 | 9054171 | ||
| BC150801.1 | - | 3 | 312 | 9087712 | ||

Ggnbp2
Genomic Map