siRNA Details
| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144827.4 | NP_659076.2 | 9 | 1102 | ||
| GenBank mRNA | BC050788.1 | - | 9 | 1102 | 6706609 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata20| Gene Symbol: Spata20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144827.4 | NP_659076.2 | 9 | 1102 | ||
| GenBank mRNA | BC050788.1 | - | 9 | 1102 | 6706609 | |