siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145828.3 | NP_665827.2 | 10 | 2206 | ||
| GenBank mRNA | AJ291751.2 | - | 10 | 2123 | ||
| AK083482.1 | - | 10 | 2176 | |||
| AK086804.1 | - | 10 | 2139 | |||
| AK150521.1 | - | 10 | 2159 | |||
| AK153332.1 | - | 10 | 2159 | |||
| BC034082.1 | - | 9 | 1725 | 4239646 | ||

Xylt2
Genomic Map