siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145828.3 | NP_665827.2 | 5, 6 | 1172 | ||
| GenBank mRNA | AJ291751.2 | - | 5, 6 | 1089 | ||
| AK083482.1 | - | 5, 6 | 1141 | |||
| AK086804.1 | - | 5, 6 | 1104 | |||
| AK150521.1 | - | 5, 6 | 1125 | |||
| AK153332.1 | - | 5, 6 | 1125 | |||
| AK158056.1 | - | 5, 6 | 1172 | |||
| AK177270.1 | - | 2, 3 | 190 | |||
| BC034082.1 | - | 4, 5 | 691 | 4239646 | ||

Xylt2
Genomic Map