siRNA Details
| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081220.1 | NP_001074689.1 | 1 | 286 | ||
| GenBank mRNA | AK030625.1 | - | 1 | 290 | ||
| AK038894.1 | - | 1 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr179| Gene Symbol: Gpr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081220.1 | NP_001074689.1 | 1 | 286 | ||
| GenBank mRNA | AK030625.1 | - | 1 | 290 | ||
| AK038894.1 | - | 1 | 280 | |||