siRNA Details
| Gene Symbol: Arl5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207231.1 | NP_997114.1 | 2 | 407 | ||
| GenBank mRNA | AK032519.1 | - | 2 | 164 | ||
| AK154137.1 | - | 2 | 127 | |||
| AK171194.1 | - | 2 | 418 | |||
| BC065791.1 | - | 2 | 407 | 3824354 | ||

Arl5c
Genomic Map