siRNA Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146030.2 | NP_666142.1 | 7 | 1319 | ||
| GenBank mRNA | AK154510.1 | - | 7 | 1335 | ||
| AK164128.1 | - | 7 | 1338 | |||
| AK189068.1 | - | 1 | 21 | |||
| BC020025.1 | - | 7 | 1319 | 3490992 | ||

Plekhh3
Genomic Map