siRNA Details
| Gene Symbol: Nags | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145829.2 | NP_665828.1 | 4 | 1005 | ||
| GenBank mRNA | AF462069.1 | - | 4 | 961 | ||
| AJ489814.1 | - | 4 | 931 | |||
| AK075765.1 | - | 3 | 651 | |||
| BC036161.1 | - | 2 | 1391 | |||
| BC057990.1 | - | 4 | 969 | 5100021 | ||

Nags
Genomic Map