siRNA Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199200.2 | NP_954670.2 | 5, 6 | 931 | ||
| GenBank mRNA | BC025575.1 | - | 5, 6 | 907 | ||
| BC064455.1 | - | 2, 3 | 664 | 6810415 | ||
| CB182176.1 | - | 4, 5 | 679 | |||
| CB202938.1 | - | 3, 4 | 397 | |||

Fam171a2
Genomic Map