siRNA Details
| Gene Symbol: Abca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147220.2 | NP_671753.2 | 20, 21 | 3023 | ||
| GenBank mRNA | AF491299.1 | - | 20, 21 | 3023 | ||
| AK029256.1 | - | 6, 7 | 812 | |||
| AK085555.1 | - | 20, 21 | 2930 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca9| Gene Symbol: Abca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147220.2 | NP_671753.2 | 20, 21 | 3023 | ||
| GenBank mRNA | AF491299.1 | - | 20, 21 | 3023 | ||
| AK029256.1 | - | 6, 7 | 812 | |||
| AK085555.1 | - | 20, 21 | 2930 | |||