siRNA Details
| Gene Symbol: PGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002630.3 | NP_002621.1 | 7, 8 | 975 | ||
| GenBank mRNA | BC073740.1 | - | 7, 8 | 957 | 6296399 | |
| J04443.1 | - | 7, 8 | 915 | |||
| U75272.1 | - | 7, 8 | 958 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGC| Gene Symbol: PGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002630.3 | NP_002621.1 | 7, 8 | 975 | ||
| GenBank mRNA | BC073740.1 | - | 7, 8 | 957 | 6296399 | |
| J04443.1 | - | 7, 8 | 915 | |||
| U75272.1 | - | 7, 8 | 958 | |||