siRNA Details
| Gene Symbol: Myo15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173262.1 | - | 12 | 1331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo15b| Gene Symbol: Myo15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173262.1 | - | 12 | 1331 | ||