siRNA Details
| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027258.1 | NP_081534.1 | 6 | 822 | ||
| GenBank mRNA | AK030994.1 | - | 2 | 61 | ||
| AK088504.1 | - | 11 | ||||
| AK171736.1 | - | 1 | 44 | |||
| BC053070.1 | - | 6 | 601 | |||
| CF539215.1 | - | 5 | 464 | |||

Rnf157
Genomic Map