siRNA Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167680.1 | NP_001161152.1 | 4 | 480 | ||
| NM_172572.3 | NP_766160.2 | 4 | 694 | |||
| GenBank mRNA | AK028872.1 | - | 4 | 695 | ||
| AK042365.1 | - | 4 | 476 | |||
| AK154639.1 | - | 4 | 449 | |||
| AK171085.1 | - | 4 | 476 | |||
| AY167043.1 | - | 4 | 480 | |||
| BC052182.1 | - | 4 | 428 | 6335022 | ||

Rhbdf2
Genomic Map