siRNA Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172573.2 | NP_766161.1 | 2, 3 | 221 | ||
| GenBank mRNA | AK048667.1 | - | 2, 3 | 223 | ||
| AK084254.1 | - | 2, 3 | 354 | |||
| AK167142.1 | - | 2, 3 | 193 | |||
| BC120924.1 | - | 2, 3 | 223 | 40097903 | ||

Engase
Genomic Map