siRNA Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139147.3 | NP_631886.2 | 1, 2 | 257 | ||
| GenBank mRNA | AB232638.1 | - | 1, 2 | 145 | ||
| AF425643.1 | - | 1, 2 | 145 | |||
| BC096601.1 | - | 1, 2 | 251 | 4974190 | ||
| BY138215.1 | - | 1, 2 | 198 | |||
| BY261509.1 | - | 1, 2 | 257 | |||

Rab40b
Genomic Map