siRNA Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139147.3 | NP_631886.2 | 4 | 430 | ||
| GenBank mRNA | AA718654.1 | - | 1 | 19 | ||
| AB232638.1 | - | 4 | 318 | |||
| AF425643.1 | - | 4 | 318 | |||
| BB847804.1 | - | 4 | 347 | |||
| BC096601.1 | - | 4 | 424 | 4974190 | ||
| BY283181.1 | - | 4 | 310 | |||

Rab40b
Genomic Map