siRNA Details
| Gene Symbol: Trib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144551.5 | NP_653134.2 | 2 | 1829 | ||
| GenBank mRNA | AF358867.1 | - | 2 | 694 | ||
| AK044747.1 | - | 2 | 1787 | |||
| AK080064.1 | - | 2 | 1032 | |||
| AK082329.1 | - | 2 | 1041 | |||
| AK144192.1 | - | 2 | 1894 | |||
| BC027159.1 | - | 1 | 286 | |||
| BC034338.1 | - | 2 | 863 | 5043179 | ||
| BC037387.1 | - | 2 | 1773 | 5363174 | ||

Trib2
Genomic Map