siRNA Details
| Gene Symbol: PGGT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005023.3 | NP_005014.2 | 7 | 759 | ||
| XM_005272020.3 | XP_005272077.2 | 7 | 774 | |||
| XM_011543490.2 | XP_011541792.1 | 7 | 774 | |||
| XM_011543492.2 | XP_011541794.1 | 7 | 771 | |||

PGGT1B
Genomic Map