siRNA Details
| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161410.1 | NP_001154882.1 | 10 | 2121 | ||
| NM_178811.4 | NP_848926.3 | 10 | 2347 | |||
| GenBank mRNA | AK049172.1 | - | 9 | 1381 | ||
| AK132166.1 | - | 10 | 2133 | |||
| AK147985.1 | - | 10 | 2121 | |||
| BC002235.1 | - | 2 | 105 | |||
| BC024077.1 | - | 10 | 2290 | 5097580 | ||
| BC030887.1 | - | 10 | 2115 | 4483611 | ||

Trappc12
Genomic Map