siRNA Details
| Gene Symbol: Snx13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014973.2 | NP_001014973.2 | 23, 24 | 2647 | ||
| GenBank mRNA | AK122352.1 | - | 12, 13 | 1296 | ||
| AK133353.1 | - | 20, 21 | 2187 | |||
| AK164161.1 | - | 23, 24 | 2572 | |||
| BC056394.1 | - | 23, 24 | 2619 | 6852346 | ||

Snx13
Genomic Map