siRNA Details
Gene Symbol: Stxbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144552.3 | NP_653135.2 | 3 | 595 | ||
GenBank mRNA | AK158779.1 | - | 3 | 481 | ||
BC024598.1 | - | 3 | 477 | 4020199 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
