siRNA Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015099.2 | NP_001015099.2 | 14, 15 | 2075 | ||
| NM_001167963.1 | NP_001161435.1 | 15, 16 | 2144 | |||
| NM_001167964.1 | NP_001161436.1 | 13, 14 | 1793 | |||
| GenBank mRNA | AK031333.1 | - | 13, 14 | 1653 | ||
| AK033513.1 | - | 15, 16 | 2088 | |||
| AK129332.1 | - | 14, 15 | 1978 | |||
| AK138847.1 | - | 1, 2 | 1074 | |||
| BC086455.1 | - | 14, 15 | 1948 | 30458160 | ||

G2e3
Genomic Map