siRNA Details
| Gene Symbol: Mis18bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172578.2 | NP_766166.2 | 8 | 2305 | ||
| GenBank mRNA | AK032698.1 | - | 6 | 1337 | ||
| AK035117.1 | - | 8 | 2304 | |||
| AK220223.1 | - | 7 | 1571 | |||
| BC052175.1 | - | 8 | 2305 | 6394068 | ||

Mis18bp1
Genomic Map