siRNA Details
| Gene Symbol: Mis18bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172578.2 | NP_766166.2 | 3 | 923 | ||
| GenBank mRNA | AK035117.1 | - | 3 | 925 | ||
| AK036395.1 | - | 3 | 933 | |||
| AK053496.1 | - | 3 | 933 | |||
| AK162657.1 | - | 3 | 947 | |||
| AK164473.1 | - | 2 | 295 | |||
| AK220223.1 | - | 2 | 189 | |||
| BC042728.1 | - | 3 | 941 | |||
| BC052175.1 | - | 3 | 923 | 6394068 | ||

Mis18bp1
Genomic Map