siRNA Details
| Gene Symbol: L2hgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145443.2 | NP_663418.1 | 3, 4 | 493 | ||
| GenBank mRNA | AK145228.1 | - | 3, 4 | 492 | ||
| AK152450.1 | - | 3, 4 | 470 | |||
| AK168429.1 | - | 3, 4 | 441 | |||
| BC016226.1 | - | 3, 4 | 437 | 4487011 | ||

L2hgdh
Genomic Map