siRNA Details
| Gene Symbol: Susd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242419.1 | NP_001229348.1 | 6 | 1023 | ||
| NM_178682.4 | NP_848797.1 | 5 | 960 | |||
| GenBank mRNA | AK030233.1 | - | 6 | 1046 | ||
| AK054203.1 | - | 4 | 675 | |||
| AK129100.1 | - | 6 | 786 | |||
| AK143867.1 | - | 6 | 1004 | |||
| AK151939.1 | - | 5 | 927 | |||
| AK152820.1 | - | 5 | 927 | |||
| AK161132.1 | - | 5 | 962 | |||
| AK164823.1 | - | 5 | 927 | |||
| BC040401.1 | - | 5 | 933 | 4193723 | ||

Susd6
Genomic Map