siRNA Details
| Gene Symbol: Mlh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029786.1 | - | 1 | 1569 | ||
| AK132195.1 | - | 2 | 3073 | |||
| AK162464.1 | - | 1 | 1740 | |||
| BC072599.1 | - | 2 | 3084 | |||
| BC079861.1 | - | 2 | 3071 | 6834767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlh3| Gene Symbol: Mlh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029786.1 | - | 1 | 1569 | ||
| AK132195.1 | - | 2 | 3073 | |||
| AK162464.1 | - | 1 | 1740 | |||
| BC072599.1 | - | 2 | 3084 | |||
| BC079861.1 | - | 2 | 3071 | 6834767 | ||