siRNA Details
| Gene Symbol: Pomt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153415.3 | NP_700464.2 | 14 | 2000 | ||
| GenBank mRNA | AF246235.1 | - | 14 | 1604 | ||
| AK043573.1 | - | 14 | 1701 | |||
| AK043713.1 | - | 14 | 1542 | |||
| AK050915.1 | - | 14 | 1569 | |||
| AK076813.1 | - | 14 | 2003 | |||
| AK214879.1 | - | 2 | 121 | |||
| AY090481.1 | - | 14 | 2000 | |||
| AY090482.1 | - | 14 | 1600 | |||
| BC052045.1 | - | 14 | 1572 | 6402328 | ||
| BC062965.1 | - | 14 | 1572 | |||
| BC079589.1 | - | 14 | 1570 | |||

Pomt2
Genomic Map