siRNA Details
| Gene Symbol: Nrde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290303.1 | NP_001277232.1 | 13 | 3405 | ||
| NM_183155.3 | NP_898978.2 | 13 | 3390 | |||
| GenBank mRNA | AW989733.1 | - | 2 | 91 | ||
| BC025577.1 | - | 8 | 2010 | 5323598 | ||
| BC051175.1 | - | 12 | 3295 | |||

Nrde2
Genomic Map