siRNA Details
| Gene Symbol: 9030617O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145448.4 | NP_663423.2 | 9 | 1425 | ||
| GenBank mRNA | AK033551.1 | - | 9 | 1454 | ||
| AK038436.1 | - | 9 | 1427 | |||
| AK081256.1 | - | 1462 | ||||
| AK152622.1 | - | 9 | 1454 | |||
| BC021385.1 | - | 10 | 1410 | 5064508 | ||
| BC066161.1 | - | 10 | 1499 | 30093441 | ||

9030617O03Rik
Genomic Map