siRNA Details
Gene Symbol: Ifi27l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145449.2 | NP_663424.1 | 4 | 281 | ||
GenBank mRNA | BC021795.1 | - | 4 | 281 | 4217067 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifi27l2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145449.2 | NP_663424.1 | 4 | 281 | ||
GenBank mRNA | BC021795.1 | - | 4 | 281 | 4217067 |