siRNA Details
| Gene Symbol: PGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000926.4 | NP_000917.3 | 3, 4 | 2647 | ||
| NM_001202474.3 | NP_001189403.1 | 3, 4 | 1897 | |||
| NM_001271162.1 | NP_001258091.1 | 3, 4 | 567 | |||
| NR_073141.2 | 3, 4 | 1897 | ||||
| XM_006718858.3 | XP_006718921.1 | 3, 4 | 3497 | |||
| GenBank mRNA | AB085845.1 | - | 3, 4 | 1904 | ||
| AF016381.1 | - | 3, 4 | 2079 | |||
| AK304853.1 | - | 3, 4 | 567 | |||
| AY382151.1 | - | 2, 3 | 263 | |||
| M15716.1 | - | 3, 4 | 2079 | |||
| X51730.1 | - | 3, 4 | 3358 | |||

PGR
Genomic Map