siRNA Details
| Gene Symbol: Rd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127685.1 | NP_001121157.1 | 2 | 170 | ||
| GenBank mRNA | AK080710.1 | - | 2 | 522 | ||
| BC048943.1 | - | 3 | 426 | |||
| BC147034.1 | - | 2 | 170 | 9056031 | ||
| BC147035.1 | - | 2 | 170 | 9056043 | ||
| BC147747.1 | - | 2 | 170 | 9056696 | ||
| BC147749.1 | - | 2 | 170 | 9056698 | ||

Rd3l
Genomic Map