siRNA Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024602.3 | NP_001019773.2 | 12 | 2718 | ||
| GenBank mRNA | AK122236.1 | - | 13 | 2647 | ||
| AK147410.1 | - | 12 | 2659 | |||
| AK147612.1 | - | 10 | 2323 | |||
| BC141174.1 | - | 11 | 2518 | 9055786 | ||

Cep170b
Genomic Map