siRNA Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146039.3 | NP_666151.3 | 19, 20 | 2633 | ||
| GenBank mRNA | AK052482.1 | - | 19, 20 | 2597 | ||
| AK077372.1 | - | 9, 10 | 1052 | |||
| AK144138.1 | - | 19, 20 | 2633 | |||
| AK144444.1 | - | 1, 2 | 187 | |||
| BC021371.1 | - | 9, 10 | 876 | |||

Wdr60
Genomic Map