siRNA Details
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 28, 29 | 4070 | ||
| GenBank mRNA | BY018585.1 | - | 1, 2 | 113 | ||
| CD563882.1 | - | 4, 5 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr1| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 28, 29 | 4070 | ||
| GenBank mRNA | BY018585.1 | - | 1, 2 | 113 | ||
| CD563882.1 | - | 4, 5 | 476 | |||