siRNA Details
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 23 | 3444 | ||
| GenBank mRNA | AK143958.1 | - | 23 | 3422 | ||
| CF895720.1 | - | 2 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr1| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 23 | 3444 | ||
| GenBank mRNA | AK143958.1 | - | 23 | 3422 | ||
| CF895720.1 | - | 2 | 377 | |||