siRNA Details
| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289546.1 | NP_001276475.1 | 20 | 1982 | ||
| NM_175007.2 | NP_778172.1 | 20 | 1970 | |||
| GenBank mRNA | AK132168.1 | - | 20 | 1983 | ||
| BC024817.1 | - | 10 | 951 | |||
| BC043718.1 | - | 4 | 639 | |||
| BC051425.1 | - | 6 | 3292 | |||
| BC054718.1 | - | 20 | 1950 | 5686478 | ||

Amph
Genomic Map