siRNA Details
| Gene Symbol: Zfp322a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111107.2 | NP_001104577.1 | 5 | 1577 | ||
| NM_001286155.1 | NP_001273084.1 | 5 | 1855 | |||
| NM_001286156.1 | NP_001273085.1 | 4 | 1780 | |||
| NM_172586.4 | NP_766174.1 | 4 | 1502 | |||
| GenBank mRNA | AK036302.1 | - | 4 | 1503 | ||
| AK036877.1 | - | 4 | 1503 | |||
| AK162608.1 | - | 4 | 1480 | |||
| AK170525.1 | - | 4 | 1497 | |||
| BC017143.1 | - | 1 | 1066 | |||
| BC032268.1 | - | 1 | 1136 | |||
| BC043711.1 | - | 4 | 1464 | 5369339 | ||

Zfp322a
Genomic Map