siRNA Details
| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146041.2 | NP_666153.1 | 2 | 187 | ||
| GenBank mRNA | AK086076.1 | - | 2 | 229 | ||
| BC031788.1 | - | 2 | 187 | 4165509 | ||
| BC093502.1 | - | 2 | 211 | 4215865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmds| Gene Symbol: Gmds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146041.2 | NP_666153.1 | 2 | 187 | ||
| GenBank mRNA | AK086076.1 | - | 2 | 229 | ||
| BC031788.1 | - | 2 | 187 | 4165509 | ||
| BC093502.1 | - | 2 | 211 | 4215865 | ||