siRNA Details
| Gene Symbol: Rnf144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170643.1 | NP_001164114.1 | 3 | 354 | ||
| NM_146042.4 | NP_666154.3 | 3 | 390 | |||
| GenBank mRNA | AK040939.1 | - | 3 | 346 | ||
| AK052846.1 | - | 3 | 355 | |||
| AK053529.1 | - | 3 | 390 | |||
| BB661852.1 | - | 3 | 355 | |||
| BC025007.1 | - | 3 | 336 | 5050848 | ||

Rnf144b
Genomic Map