siRNA Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033268.2 | NP_001028440.2 | 15 | 2704 | ||
| GenBank mRNA | AK035074.1 | - | 5 | 619 | ||
| AK172909.1 | - | 15 | 2893 | |||
| BC010304.1 | - | 11 | 1772 | |||
| BC033412.1 | - | 7 | 1032 | |||
| BC042582.1 | - | 14 | 2220 | |||
| BC158134.1 | - | 15 | 2696 | |||

Fam120a
Genomic Map