siRNA Details
| Gene Symbol: Zfp455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048204.1 | NP_001041669.1 | 4 | 1429 | ||
| GenBank mRNA | AK028336.1 | - | 3 | 1274 | ||
| BC010651.1 | - | 1 | 876 | |||
| Gene Symbol: Zfp456 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001186.3 | NP_001001186.3 | 4 | 1414 | ||
| GenBank mRNA | AK164227.1 | - | 4 | 1416 | ||
| BC141197.1 | - | 4 | 1397 | 9055809 | ||

Zfp455
Genomic Map