siRNA Details
| Gene Symbol: Zfp595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177622.3 | NP_808290.1 | 4 | 1123 | ||
| GenBank mRNA | AK038563.1 | - | 4 | 1124 | ||
| AK134150.1 | - | 4 | 1101 | |||
| AK136304.1 | - | 4 | 1111 | |||
| AK154058.1 | - | 4 | 1101 | |||
| BC141018.1 | - | 4 | 1196 | 9055630 | ||

Zfp595
Genomic Map