siRNA Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131068.1 | NP_001124540.1 | 4 | 875 | ||
| NM_001131069.1 | NP_001124541.1 | 4 | 338 | |||
| NM_178916.5 | NP_849247.3 | 4 | 1325 | |||
| GenBank mRNA | AK029602.1 | - | 4 | 1151 | ||
| AK040255.1 | - | 4 | 830 | |||
| AK083705.1 | - | 4 | 1326 | |||
| BC030381.1 | - | 4 | 950 | 5375753 | ||
| CD769672.1 | - | 4 | 342 | |||
| CN839985.1 | - | 4 | 348 | |||

Rfesd
Genomic Map