siRNA Details
| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081352.1 | NP_001074821.1 | 43 | 4953 | ||
| GenBank mRNA | AK129128.1 | - | 22 | 2309 | ||
| AK163406.1 | - | 3 | 305 | |||
| BC027390.1 | - | 5 | 543 | |||
| BC033466.1 | - | 5 | 543 | |||
| BC035338.1 | - | 24 | 3241 | |||
| EF591876.1 | - | 4442 | ||||

Ttc37
Genomic Map