siRNA Details
| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081352.1 | NP_001074821.1 | 38 | 4243 | ||
| GenBank mRNA | AK129128.1 | - | 17 | 1599 | ||
| BC035338.1 | - | 19 | 2531 | |||
| EF591876.1 | - | 3748 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc37| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081352.1 | NP_001074821.1 | 38 | 4243 | ||
| GenBank mRNA | AK129128.1 | - | 17 | 1599 | ||
| BC035338.1 | - | 19 | 2531 | |||
| EF591876.1 | - | 3748 | ||||