siRNA Details
| Gene Symbol: Ankrd34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175455.4 | NP_780664.2 | 5 | 1607 | ||
| GenBank mRNA | AK078203.1 | - | 5 | 1681 | ||
| AK090387.1 | - | 5 | 1611 | |||
| AK138465.1 | - | 4 | 1793 | |||
| BC096609.1 | - | 5 | 1482 | |||
| BC119312.1 | - | 5 | 1469 | 8734061 | ||
| BC119314.1 | - | 5 | 1469 | 8734063 | ||

Ankrd34b
Genomic Map