siRNA Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173392.4 | NP_775568.1 | 3 | 409 | ||
| GenBank mRNA | AK045090.1 | - | 3 | 467 | ||
| AK122241.1 | - | 3 | 390 | |||
| AK170435.1 | - | 3 | 428 | |||
| BC042669.1 | - | 3 | 407 | 4235024 | ||

Zfyve16
Genomic Map